Would you like to log in again?
Tech-Med® Stainless Steel Bowls, DUKAL Corporation®
Supplier: Dukal
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4252 4250 4251 4247 4248
76235-152BX 464.95 USD InStock
76235-152 76235-150 76235-156 76235-154 76235-158
Tech-Med® Stainless Steel Bowls, DUKAL Corporation®
Trays/Dishes/Pots Laboratory Trays/Dishes
Bowls are crafted without seams to prevent bacteria growth and allow for easy cleaning.
  • Made of the highest grade stainless steel (18/8)
  • Autoclavable
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty