Would you like to log in again?
Tech-Med® Stainless Steel Tumblers, DUKAL
Supplier: Dukal
4242 4242-1
76235-142BX 155.18 USD InStock
76235-142 76235-468
Tech-Med® Stainless Steel Tumblers, DUKAL
Beakers
Made of the high grade stainless steel (18/8, 304).
  • Crafted without seams which prevents bacteria growth and allows for easy cleaning
  • Autoclavable
  • Non sterile
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty