Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT3 Mouse Monoclonal Antibody [Clone: 9A5.H9.G7]

Supplier: Rockland Immunochemicals
KCAE75
76235-516EA 426 USD InStock
76235-516
Anti-AKT3 Mouse Monoclonal Antibody [Clone: 9A5.H9.G7]
Antibodies
AKT3 Western Bloting kit allows for the detection of endogenous protein levels of human AKT3 present in cell lysates provided by the user. As a positive control, this kit includes a MDA-MB468 whole cell lysate proven to contain AKT3. After protein separation by SDS-PAGE and transfer, the membrane is probed with Rockland's optimized Anti-AKT3 antibody. Detection of the membrane bound antibody-antigen complex is achieved by the addition of a secondary antibody conjugated to the enzyme horseradish peroxidase. The enzyme reacts with a specialized formulation of luminol, an extremely sensitive, non-radioactive substrate that emits light and allows visualization using X-ray film or other imaging methods, including highly sensitive CCD cameras and imaging systems. AKT3 Western Bloting is ideal for investigators involved in Cell Signaling, Cancer, Neuroscience and Signal Transduction research.

Use Rockland Immunochemicals’ Anti-AKT3 Chemiluminescent Kit for Western Blotting for detection of human AKT3 native and recombinant proteins by western blot. Expect a band approximately 56 kDa in size corresponding to AKT3 protein. This kit is useful for both western blotting and dot blotting methods. Please read the entire product insert prior to use.

Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 9A5.H9.G7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty