Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
K-GM8
76235-538EA 158 USD InStock
76235-538
Anti-PARP1 Rabbit Polyclonal Antibody
Antibodies
PARP1 is the primary member of the poly(ADP-ribose) polymerase family, whose function is to signal DNA damage (and to recruit repair proteins) by PARylation. PARP1 is also involved in multiple cell death pathways, including apoptosis, necroptosis, autophagy, and a relatively new pathway termed parthanatos. It has been implicated in a new form of cell death termed parthanatos. PARP1 can also promote tissue survival by shifting the balance of cell death programs between autophagy and necrosis. Clinical studies have shown vulnerability to PARP inhibitors in DNA repair defective cancers. Anti-PARP1 (N-term ZF1) antibody is useful for researchers interested in cellular processes including DNA damage, transcriptional control, and stem cell identity research.

Anti-PARP1 (N-term ZF1) antibody with the matched secondary has been validated by western blotting and nanoimmunoassay (NIA). Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 113 kDa in size corresponding to PARP-1 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: PARP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty