Would you like to log in again?
Human CXCL9 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0427
76235-588EA 687 USD InStock
76235-588
Human CXCL9 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CXCL9 AccuSignal ELISA Kit
  • Natural and recombinant human CXCL9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: T23-T125
Chemokine(C-X-C motif) ligand 9(CXCL9) is a small cytokine belonging to the CXC chemokine family that is also known as Monokine induced by gamma interferon(MIG). CXCL9 is a T-cell chemoattractant, which is induced by IFN-gamma. It is closely related to two other CXC chemokines called CXCL10 and CXCL11, whose genes are located near the gene for CXCL9 on human chromosome 4.1, 2 CXCL9, CXCL10 and CXCL11 all elicit their chemotactic functions by interacting with the chemokine receptor CXCR3.3 The standard product used in this kit is recombinant human CXCL9, consisting of 103 amino acids with the molecular mass of 11.7KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human CXCL9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CXCL9 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty