Would you like to log in again?
Human HBEGF ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0456
76235-630EA 636 USD InStock
76235-630
Human HBEGF ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human HBEGF AccuSignal ELISA Kit
  • Natural and recombinant human HBEGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: D63-L148
HBEGF(Heparin-binding EGF-like growth factor), also known as HEGFL or DTR, is a member of the EGF family of proteins that in humans is encoded by the HBEGF gene. The HBEGF gene is assigned to chromosome 5, thus confirming the assignment of the gene on the basis of its role in relation to diphtheria toxin susceptibility. HB-EGF is an 87 amino acid glycoprotein which displays highly regulated gene expression. It has been shown to play a role in wound healing, cardiac hypertrophy and heart development and function. HB-EGF binding and activation of EGF receptors plays a critical role during cardiac valve tissue development and the maintenance of normal heart function in adults.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human HBEGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human HBEGF standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty