Would you like to log in again?
Human IL-6R alpha ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0458
76235-634EA 636 USD InStock
76235-634
Human IL-6R alpha ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-6R alpha AccuSignal ELISA Kit
  • Natural and recombinant human IL-6R alpha.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: L20-D358
IL6R alpha(IL6RA) also known as CD126 or IL6R, is a type I cytokine receptor. The IL6 receptor is a protein complex consisting of a IL-6 receptor subunit(IL6R) and interleukin 6 signal transducer Glycoprotein 130. IL6R also denotes the human gene encoding this subunit. Alternatively spliced transcript variants encoding distinct isoforms have been reported. IL6R subunit is also shared by many other cytokines. Interleukin-6 receptor has been shown to interact with Interleukin 6 and Ciliary neurotrophic factor. Ligand binding did not appear to affect IL6R dimerization status. IL6R dimerization occurs both on the cell surface and in solution.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human IL-6R alpha standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA, citrate) or urine to each empty well. It is recommended that each human IL-6R alpha standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty