Would you like to log in again?
Human IL-7 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0459
76235-636EA 636 USD InStock
76235-636
Human IL-7 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-7 AccuSignal ELISA Kit
  • Natural and recombinant human IL-7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: D26-H177
IL-7 a hematopoietic growth factor secreted by stromal cells in the red marrow and thymus. It is also produced by keratinocytes,1 dendritic cells,2 hepatocytes,3 neurons, and endothelial cells4. Interleukin 7(IL7) is a protein5 that in humans is encoded by the IL7 gene.6, 7, 8Knockout studies in mice suggested that this cytokine plays an essential role in lymphoid cell survival.9 IL-7 is a cytokine important for B and T cell development. This cytokine and the hepatocyte growth factor(HGF) form a heterodimer that functions as a pre-pro-B cell growth-stimulating factor. This cytokine is found to be a cofactor for V(D)J rearrangement of the T cell receptor beta(TCRB) during early T cell development.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human IL-7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(EDTA) to each empty well. It is recommended that each human IL-7 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty