Would you like to log in again?
Human FLRG - FSTL3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0526
76235-648EA 687 USD InStock
76235-648
Human FLRG - FSTL3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human FLRG - FSTL3 AccuSignal ELISA Kit
  • Natural and recombinant human FLRG.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M27-V263
FSTL3(Follistatin-Like 3) also known as FLRG or FOLLISTATIN-RELATED GENE, is a member of the follistatin-module protein family, which is composed of extracellular matrix-associated glycoproteins thought to act in a paracrine manner to bind morphogens or growth/differentiation factors and regulate their activity during development. The FSTL3 gene extends over 7 kbp and contains 5 exons. The FSTL3 gene was localized to chromosome 19p13. This gene involved in hematopoiesis and in differentiation of hemopoietic progenitor cells, increases hematopoietic cell adhesion to fibronectin and seems to contribute to the adhesion of hematopoietic precursor cells to the bone marrow stroma. It may have a role in leukemogenesis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human FLRG standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), urine or human milk to each empty well. It is recommended that each human FLRG standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty