Would you like to log in again?
Human PECAM-1 - CD31 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0527
76235-650EA 713 USD InStock
76235-650
Human PECAM-1 - CD31 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human PECAM-1 - CD31 AccuSignal ELISA Kit
  • Natural and recombinant human PECAM-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: cho; Immunogen sequence: Q28-K601
Human platelet/endothelial cell adhesion molecule-1(PECAM1), an important member of the immunoglobulin gene superfamily, is widely distributed on cells of the vascular system and mediates cellular interactions through both homophilic and heterophilic adhesive mechanisms. The function of PECAM1 in vitro has begun to be understood, but its function in vivo is yet to be established.1 The PECAM1 locus is on the long arm of chromosome 17, in the region q23-qter. To confirm this observation and obtain a more precise localization of the PECAM1 locus, fluorescence in situ hybridization was conducted. Together our data allowed assignment of the PECAM1 locus to the region 17q23.2 To examine the functional role of PECAM-1 in regulating platelet-collagen interactions, 2 different approaches were applied using recombinant human PECAM-1-immunoglobulin chimeras and platelets derived from PECAM-1-deficient mice.3 PECAM-1, an integral membrane protein with an essential role in TEM, is found in this compartment and constitutively recycles evenly along endothelial cell borders.4 The standard used in this kit is the product of gene recombinant expression(extracellular part), consisting of 574 amino acids(Q28-K601) with the molecular weight of 64.5KDa. After glycosylating, its molecular weight changes to 90-100KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human PECAM-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human PECAM-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty