Would you like to log in again?
Mouse KIM1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0529
76235-654EA 652 USD InStock
76235-654
Mouse KIM1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse KIM1 AccuSignal ELISA Kit
  • Natural and recombinant mouse KIM1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Y22-T212
KIM1(TIM-1), also known as Hepatitis A virus cellular receptor 1, is a protein that in Mouses is encoded by the HAVCR1 gene.[1][2][3] Infection of canine osteogenic sarcoma cells expressing HAVCR1 with HAV led Feigelstock et al.(1998) to conclude that the protein is indeed a receptor for the virus. Immunofluorescence microscopy demonstrated internalization of HAV by dog cells expressing HAVCR1. Using a monoclonal antibody to mouse Tim1, Umetsu et al.(2005) showed that Tim1 was expressed after activation of naive T cells and on T cells differentiated in Th2-polarizing conditions. By homology of synteny with the mouse Tim1 gene and database analysis, McIntire et al.(2001) mapped the HAVCR1 gene to 5q33.2. The standard used in this kit is recombinant mouse KIM-1, constituting from Y22-T212 amino acids, and 191amino acids with the molecular mass of 21.8KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse KIM1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, tissue homogenates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each mouse KIM1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty