Would you like to log in again?
Human Tryptase - TPSAB1,B2 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0539
76235-674EA 656 USD InStock
76235-674
Human Tryptase - TPSAB1,B2 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Tryptase - TPSAB1,B2 AccuSignal ELISA Kit
  • Natural and recombinant human Tryptase.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-V275
Thrombospondin 1, also known as THBS1, is a protein that in humans in encoded by the THBS1 gene. Thrombospondin 1 is a subunit of a disulfide-linked homotrimeric protein. This protein is an adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. By in situ hybridization, the THBS1 gene was mapped to human 15q15 and the cognate gene to mouse chromosome 2(region F) and was localized to 15q11-qter by Southern analysis of human-rodent somatic cell hybrids. Thrombospondin I is a multimodular secreted protein that associates with the extracellular matrix and possesses a variety of biologic functions, including a potent antiangiogenic activity. Other thrombospondin genes include thrombospondins II, III, and IV.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Tryptase standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Tryptase standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty