Would you like to log in again?
Rat CCL4 - MIP-1 beta ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0574
76235-688EA 636 USD InStock
76235-688
Rat CCL4 - MIP-1 beta ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat CCL4 - MIP-1 beta AccuSignal ELISA Kit
  • Natural and recombinant rat CCL4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A24-N92
Chemokine(C-C motif) ligand 4, also known as CCL4, is a protein which in mouse is encoded by the CCL4 gene. It is a CC chemokine with specificity for CCR5 receptors. It is a chemoattractant for natural killer cells, monocytes and a variety of other immune cells. CCL4 is a major HIV-suppressive factor produced by CD8+ T cells. Performing-low memory CD8+ T cells that normally synthesize MIP-1-beta. By analysis of somatic cell hybrids and by in situ hybridization, the CCL4 gene was assigned to 17q21-q23. Based on functional analysis of MIP-1beta, one or more additional beta chemokine receptors that transduce MIP-1beta signals must also exist. MIP-1alpha and MIP-1beta have been shown to lack affinity for the erythrocyte surface chemokine receptor/Duffy antigen that binds many alpha as well as beta chemokines(11, 14).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat CCL4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each rat CCL4 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty