Would you like to log in again?
Human MIA ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0576
76235-692EA 636 USD InStock
76235-692
Human MIA ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human MIA AccuSignal ELISA Kit
  • Natural and recombinant human MIA.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: G25-131Q
Melanoma Inhibiting Activity(MIA), also known as cartilage-derived retinoic acid-sensitive protein(CD-RAP), is an approximately 11-15 kDa protein that is expressed as a noncovalent homodimer. MIA is structurally related to OTOR/Otoraplin and MIA-2 in a small family of secreted proteins with one SH3 domain(1-3). And the MIA gene was mapped to 19q13.32-q13.33 by fluorescence in situ hybridization. Beside, It is a marker for malignant melanoma. MIA functions as a chemoattractant for mesenchymal stem cells and enhances their BMP-2 and TGF-beta3 induced differentiation into chondrocytes . MIA-deficient mice exhibit delayed chondrocyte differentiation but enhanced chondrocyte proliferation and cartilage repair .
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human MIA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or saliva to each empty well. It is recommended that each human MIA standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty