Would you like to log in again?
Human PD-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0589
76235-716EA 687 USD InStock
76235-716
Human PD-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human PD-1 AccuSignal ELISA Kit
  • Natural and recombinant human PD-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L25-Q167
Programmed cell death protein 1, also called PD-1 or PDCD1 is a protein that in humans is encoded by the PDCD1 gene. This gene encodes a cell surface membrane protein of the immunoglobulin superfamily. By fluorescence in situ hybridization, the PDCD1 gene is mapped to 2q37.3. This protein is expressed in pro-B-cells and is thought to play a role in their differentiation. In mice, expression of this gene is induced in the thymus when anti-CD3 antibodies are injected and large numbers of thymocytes undergo apoptosis. Mice deficient for this gene bred on a BALB/c background developed dilated cardiomyopathy and died from congestive heart failure. These studies suggest that this gene product may also be important in T cell function and contribute to the prevention of autoimmune diseases.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human PD-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates to each empty well. It is recommended that each human PD-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty