Would you like to log in again?
Human FSTL1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0595
76235-728EA 702 USD InStock
76235-728
Human FSTL1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human FSTL1 AccuSignal ELISA Kit
  • Natural and recombinant human FSTL1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-I308
Follistatin-related protein 1, also called FSTL1 or FRP is a protein that in humans is encoded by the FSTL1 gene. FSTL1 gene is mapped to chromosome 3q13.33 based on an alignment of FSTL1 sequence with the genomic sequence(GRCh37). This gene encodes a protein with similarity to follistatin, an activin-binding protein. It contains an FS module, a follistatin-like sequence containing 10 conserved cysteine residues. This gene product is thought to be an autoantigen associated with rheumatoid arthritis. This gene may modulate the action of some growth factors on cell proliferation and differentiation. It was suggested that fibroblast-secreted FSTL1 acts on macrophages to increase TNF and IL1B production and IL6 activity.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human FSTL1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma( heparin, EDTA) to each empty well. It is recommended that each human FSTL1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty