Would you like to log in again?
Human IGFBP7 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0618
76235-758EA 636 USD InStock
76235-758
Human IGFBP7 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human IGFBP7 AccuSignal ELISA Kit
  • Natural and recombinant human IGFBP7.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D30-L282
Insulin-like growth factor-binding protein 7 is a protein that in humans is encoded by the IGFBP7 gene. The gene was mapped to chromosome 4q12. The major function of the protein is the regulation of availability of insulin-like growth factors(IGFs) in tissue as well as in modulating IGF binding to its receptors. IGFBP7 binds to IGF with high affinity. It also stimulates cell adhesion. The protein is implicated in some cancers.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 40000pg/ml, 20000pg/ml, 10000pg/ml, 5000pg/ml, 2,500pg/ml, 1,250pg/ml, 625pg/ml human IGFBP7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each human IGFBP7 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty