Would you like to log in again?
Mouse IL-31 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0626
76235-774EA 687 USD InStock
76235-774
Mouse IL-31 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-31 AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-31.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: T24-C163
Interleukin-31, also known as IL-31 is a protein that in humans is encoded by the IL31 gene. The IL31 gene is mapped to chromosome 12q24.31 and the mouse gene maps to a syntenic region on chromosome 5. IL31, which is made principally by activated Th2-type T cells, interacts with a heterodimeric receptor consisting of IL31RA and OSMR that is constitutively expressed on epithelial cells and keratinocytes. IL31 may be involved in the promotion of allergic skin disorders and in regulating other allergic diseases, such as asthma.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse IL-31 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-31 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty