Would you like to log in again?
Human A2M-alpha2-Macroglobulin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0638
76235-798EA 636 USD InStock
76235-798
Human A2M-alpha2-Macroglobulin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human A2M-alpha2-Macroglobulin AccuSignal ELISA Kit
  • Natural human A2M.
  • There is no detectable cross-reactivity with other relevant proteins.
  • From human plasma
Alpha-2-macroglobulin, also known as A2M or CPAMD5 is a large plasma protein found in the blood. This gene is mapped to 12p13.31. Alpha-2-macroglobulin is a protease inhibitor and cytokine transporter. It inhibits many proteases, including trypsin, thrombin and collagenase. A2M is implicated in Alzheimer disease(AD) due to its ability to mediate the clearance and degradation of A-beta, the major component of beta-amyloid deposits. This gene is able to inhibit all four classes of proteinases by a unique 'trapping' mechanism. This protein has a peptide stretch, called the 'bait region' which contains specific cleavage sites for different proteinases. When a proteinase cleaves the bait region, a conformational change is induced in the protein which traps the proteinase. The entrapped enzyme remains active against low molecular weight substrates(activity against high molecular weight substrates is greatly reduced). Following cleavage in the bait region a thioester bond is hydrolyzed and mediates the covalent binding of the protein to the proteinase.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 40,000pg/ml, 20,000pg/ml, 10,000pg/ml, 5,000pg/ml, 2,500pg/ml, 1,250pg/ml, 625pg/ml human A2M standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human A2M standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty