Would you like to log in again?
Human TNFRSF13C - BAFFR ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0641
76235-804EA 650 USD InStock
76235-804
Human TNFRSF13C - BAFFR ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TNFRSF13C - BAFFR AccuSignal ELISA Kit
  • Natural and recombinant human TNFRSF13C.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S7-A71
Tumor necrosis factor receptor superfamily member 13C also known as B-cell-activating factor receptor or BAFF receptor is a protein that in humans is encoded by the TNFRSF13C gene. By homology to a BAC clone, the BAFFR gene was mapped to chromosome 22q13.1-q13.31. B-cell activating factor(BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. The protein encoded by this gene is a receptor for BAFF and is a type III transmembrane protein containing a single extracellular phenylalanine-rich domain. It is thought that this receptor is the principal receptor required for BAFF-mediated mature B-cell survival.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml, 78pg/ml human TNFRSF13C standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human TNFRSF13C standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty