Would you like to log in again?
Human Nesfatin-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0652
76235-818EA 636 USD InStock
76235-818
Human Nesfatin-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Nesfatin-1 AccuSignal ELISA Kit
  • Natural and recombinant mouse CCL8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: V25-L106
Nesfatin-1 is a naturally occurring protein molecule produced by the brains of mammals. It is responsible for regulating appetite and production of body fat. Nesfatin-1 is a metabolic polypeptide encoded in the N-terminal region of the protein precursor, Nucleobindin2(NUCB2). Recombinant human Nesfatin-1 is a 9.7 kDa protein containing 82 amino acid residues. Originally identified as a hypothalamic neuropeptide, Nesfatin-1 is also expressed in other areas of the brain, and in pancreatic islets of Langerhans, gastric endocrine cells and adipocytes. Nesfatin-1 plays a role in hunger and energy regulation in an independent manner.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human Nesfatin-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human Nesfatin-1 standard solution and each sample be measured in duplicate.
Seal the plate with the cover and incubate at 37°C for 90 min.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty