Would you like to log in again?
Mouse Sclerostin - SOST ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0679
76235-838EA 693 USD InStock
76235-838
Mouse Sclerostin - SOST ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Sclerostin - SOST AccuSignal ELISA Kit
  • Natural and recombinant mouse Sclerostin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q24-Y211
Sclerostin is a protein that in humans is encoded by the SOST gene. Sclerostin is a secreted glycoprotein with a C-terminal cysteine knot-like(CTCK) domain and sequence similarity to the DAN(differential screening-selected gene aberrative in neuroblastoma) family of bone morphogenetic protein(BMP) antagonists. Sclerostin is produced by the osteocyte and has anti-anabolic effects on bone formation. The SOST gene maps to chromosome 17q12-q21
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse Sclerostin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well.. It is recommended that each mouse Sclerostin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty