Would you like to log in again?
Mouse IGFBP5 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0682
76235-844EA 636 USD InStock
76235-844
Mouse IGFBP5 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse IGFBP5 AccuSignal ELISA Kit
  • Natural and recombinant mouse IGFBP5.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L20-E271
Insulin-like growth factor-binding protein 5 is a protein that in humans is encoded by the IGFBP5 gene. The IGFBP5 gene was mapped to chromosome 2q33-q34. The expression of IGFBP5 by stable transfection and adenovirus-mediated infection was inhibitory to growth in 2 human breast cancer cell lines. Stable expression of IGFBP5 in the breast cancer cell lines also inhibited the formation and growth of tumors following injection in athymic mice. IGFBP5 is believed a growth inhibitor and proapoptotic agent in breast cancer cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse IGFBP5 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IGFBP5 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty