Would you like to log in again?
Mouse SP-D ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0685
76235-850EA 636 USD InStock
76235-850
Mouse SP-D ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse SP-D AccuSignal ELISA Kit
  • Natural and recombinant mouse SP-D.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: A20-F374
Surfactant, pulmonary-associated protein D, also known as SFTPD or SP-D, is a protein which in humans is encoded by the SFTPD gene. By fluorescence in situ hybridization, the SP-D gene was localized in 10q22.2-q23.1. On the basis of homology with other collectins, potential functions for SP-D include roles in innate immunity and surfactant metabolism, SP-D is produced in the bronchiolar and terminal epithelium of human fetal lung from about 21 weeks of gestation. What's more, SP-A and SP-D act as dual-function surveillance molecules that reverse orientation and function and become initiators of host-defense reactions.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml mouse SP-D standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse SP-D standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty