Would you like to log in again?
Mouse MSP - MST1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0688
76235-856EA 636 USD InStock
76235-856
Mouse MSP - MST1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse MSP - MST1 AccuSignal ELISA Kit
  • Natural and recombinant mouse MSP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: Q19-E716
Macrophage Stimulating 1(SMT1), also known as Hepatocyte growth factor-like protein(HGFL) and MSP, is a protein that in humans is encoded by the MST1 gene. The HGFL gene was identified at the DNF15S2 locus on human chromosome 3(3p21). The RON tyrosine kinase), the receptor for MSP, is expressed on the ciliated epithelia of the mucociliary transport apparatus of the lung. Furthermore, MSP stimulated ciliary motility in these cells by activating RON.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse MSP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse MSP standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty