Would you like to log in again?
Mouse SAA ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0689
76235-858EA 636 USD InStock
76235-858
Mouse SAA ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse SAA AccuSignal ELISA Kit
  • Natural and recombinant mouse SAA.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: G20-Y122
Serum amyloid A protein(SAA), also known as SAA1, is a protein that in humans is encoded by the SAA1 gene. This gene encodes a member of the serum amyloid A family of apolipoproteins. It is mapped to 11p15.1. SAA is a major acute phase protein that is highly expressed in response to inflammation and tissue injury. This protein also plays an important role in HDL metabolism and cholesterol homeostasis. High levels of this protein are associated with chronic inflammatory diseases including atherosclerosis, rheumatoid arthritis, Alzheimer's disease and Crohn's disease. This protein may also be a potential biomarker for certain tumors.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse SAA standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each mouse SAA standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty