Would you like to log in again?
Human TGFBI - beta IG-H3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0699
76235-878EA 636 USD InStock
76235-878
Human TGFBI - beta IG-H3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TGFBI - beta IG-H3 AccuSignal ELISA Kit
  • Natural and recombinant human TGFBI.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: G24-H683
Transforming growth factor, beta-induced, 68kDa, also known as TGFBI(initially called BIGH3, BIG-H3), is a protein which in humans is encoded by the TGFBI gene. It is mapped to 5q31.1. This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human TGFBI standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human TGFBI standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty