Would you like to log in again?
Human PSP94 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0704
76235-888EA 840 USD InStock
76235-888
Human PSP94 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human PSP94 AccuSignal ELISA Kit
  • Natural and recombinant human PSP94.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S21-I114
Beta-microseminoprotein, also known as MSMB or PSP94, is a protein that in humans is encoded by the MSMB gene. It is a member of the immunoglobulin binding factor family. This gene is mapped to 10q11.23. It is synthesized by the epithelial cells of the prostate gland and secreted into the seminal plasma. The expression of the encoded protein is found to be decreased in prostate cancer. But transcripts of identical size were found in prostate, respiratory(tracheal, bronchial, and lung) tissues, and the mucosa of the gastric antrum. Thus, the protein may have functions other than those related to male reproduction. This protein has inhibin-like activity. It may have a role as an autocrine paracrine factor in uterine, breast and other female reproductive tissues.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human PSP94 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human PSP94 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty