Would you like to log in again?
Rat MIP-1 alpha - CCL3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KOA0712
76235-904EA 636 USD InStock
76235-904
Rat MIP-1 alpha - CCL3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Rat MIP-1 alpha - CCL3 AccuSignal ELISA Kit
  • Natural and recombinant rat MIP-1 alpha.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A24-A92
Macrophage inflammatory protein-1 alpha(MIP-1 alpha), also called CCL3, LD78. The cDNA for MIP-1 alpha predicts a mature peptide of 69 amino acids with a molecular mass of 7,889 daltons. LD78 is a member of a newly identified superfamily of small inducible proteins involved in inflammatory responses, wound healing and tumorigenesis. MIP-1 alpha is a chemokine that has pro-inflammatory and stem cell inhibitory activities in vitro. It constitutes an important second signal for mast cell degranulation in the conjunctiva in vivo and consequently for acute-phase disease.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml rat MIP-1α standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each rat MIP-1α standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty