Would you like to log in again?
Mouse TNFRSF14 - HVEM ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0720
76235-920EA 636 USD InStock
76235-920
Mouse TNFRSF14 - HVEM ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse TNFRSF14 - HVEM AccuSignal ELISA Kit
  • Natural and recombinant mouse TNFRSF14.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: Q29-V207
Tumor necrosis factor receptor superfamily member 14(TNFRSF14), also known as HVEM, is a protein that in humans is encoded by the TNFRSF14 gene. The protein encoded by this gene is a member of the TNF-receptor superfamily. It is mapped to 1p36.32. HVEM plays an important role in HSV pathogenesis because it enhanced the entry of several wildtype HSV strains of both serotypes into CHO cells, and mediated HSV entry into activated human T cells. HVEM and BTLA which are form a bidirectional signaling pathway can regulate cell survival and inhibitory responses between interacting cells. HVEM as an important orchestrator of mucosal immunity integrates signals from innate lymphocytes to induce optimal epithelial Stat3 activation, which indicated that targeting HVEM with agonists could improve host defense.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse TNFRSF14 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TNFRSF14 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty