Would you like to log in again?
Human TFF3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0726
76235-932EA 642 USD InStock
76235-932
Human TFF3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human TFF3 AccuSignal ELISA Kit
  • Natural and recombinant human TFF3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: E22-Y80
Trefoil factor 3 is a protein that in humans is encoded by the TFF3 gene. It is mapped to 21q22.3. Members of the trefoil family can interact with mucins and have an influence on mucus viscosity. They also promote migration of epithelial cells, are linked to antiapoptosis, induce cell scattering, trigger chemotaxis, and participate in immune responses. This gene is a marker of columnar epithelium and is expressed in a variety of tissues including goblet cells of the intestines and colon. It has been found that TFF3 is the most highly upregulated gene in 19 poorly differentiated endometrioid endometrial carcinomas (G3-EECs) compared with normal endometrium biopsies.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human TFF3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), saliva or urine to each empty well. It is recommended that each human TFF3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty