Would you like to log in again?
Human SPINK1 - TATI ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0730
76235-940EA 840 USD InStock
76235-940
Human SPINK1 - TATI ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human SPINK1 - TATI AccuSignal ELISA Kit
  • Natural and recombinant human SPINK1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: D24-C79
Serine protease inhibitor Kazal-type 1, also called TATI or PSTI, is a protein that in humans is encoded by the SPINK1 gene. It is mapped to 5q32. SPINK1 gene contains 4 exons spanning approximately 7.5 kb. It was initially detected in urine of patients with ovarian cancer. This peptide is also produced by the mucosa of the gastrointestinal tract where it may protect the mucosal cells from proteolytic breakdown. The SPINK1 gene encodes pancreatic secretory trypsin inhibitor, which is secreted from pancreatic acinar cells into pancreatic juice. Its physiologic role was thought to be the prevention of trypsin-catalyzed premature activation of zymogens within the pancreas and the pancreatic duct. Mutations in this gene are associated with hereditary pancreatitis and tropical calcific pancreatitis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human SPINK1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human SPINK1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty