Would you like to log in again?
Human GPNMB - Osteoactivin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0737
76235-946EA 636 USD InStock
76235-946
Human GPNMB - Osteoactivin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human GPNMB - Osteoactivin AccuSignal ELISA Kit
  • Natural and recombinant human GPNMB.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A22-P486
Transmembrane glycoprotein NMB is a protein that in humans is encoded by the GPNMB gene. In osteoblast progenitor cells, GPNMB works as a positive regulator of osteoblast differentiation during later stages of matrix maturation and mineralization that is mediated at least in part by BMP-2 in a SMAD1 dependent manner to promote osteoblast differentiation. GPNMB can enhance the repairing process in bone fracture, demonstrated by its high expression during chondrogenesis(soft callus) and osteogenesis(hard callus) compared to the intact femurs that is why Osteoactivin(OA) could be a novel therapeutic agent used to treat generalized osteoporosis or localized osteopenia during fracture repair by stimulating bone growth and regeneration. Similarly, GPNMB expression increases during osteoclast differentiation and it is functionally implicated in this process, possibly by promoting the fusion of osteoclast progenitor cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human GPNMB standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human GPNMB standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty