Would you like to log in again?
Human IL-18R1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0744
76235-952EA 636 USD InStock
76235-952
Human IL-18R1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-18R1 AccuSignal ELISA Kit
  • Natural and recombinant human IL-18R1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: C22-R329
The interleukin-18 receptor 1(IL-18R1), also known as CDw218a(cluster of differentiation w218a) or IL18RA, is an interleukin receptor of the immunoglobulin superfamily. The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2(IL-1Rrp2), and IL1RL1(T1/ST2), form a gene cluster on chromosome 2q. This receptor specifically binds interleukin 18(IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human IL-18R1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-18R1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty