Would you like to log in again?
Mouse Lumican ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0745
76235-954EA 636 USD InStock
76235-954
Mouse Lumican ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Lumican AccuSignal ELISA Kit
  • Natural and recombinant mouse Lumican.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q19-N338
Lumican, also known as LUM, is a protein which in humans is encoded by the LUM gene. This gene is a member of the small interstitial proteoglycan gene(SIPG) family, and it is a keratan sulfate proteoglycan which presents large quantities in the corneal stroma and in interstitial collagenous matrices of the heart, aorta, skeletal muscle, skin, and intervertebral discs. Lumican is mapped to 12q21.33, it interacts with collagen and limits growth of fibrils in diameter. In the cornea, Lumican not only interacts with collagen molecules to limit fibril growth, but also plays a critical role in the regular spacing of fibrils and acquisition of corneal transparency by virtue of its keratan sulfate-containing glycosaminoglycan side chains LDC.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse Lumican standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum or plasma(heparin) to each empty well. It is recommended that each mouse Lumican standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty