Would you like to log in again?
Mouse Prostasin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0749
76235-962EA 636 USD InStock
76235-962
Mouse Prostasin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Prostasin AccuSignal ELISA Kit
  • Natural and recombinant mouse Prostasin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A30-Q289
Prostasin is a protein that in humans is encoded by the PRSS8 gene. It is a single-copy gene and mapped to 16p11.2. Prostasin is likely to be the more physiologically relevant protease. This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. Coexpression of Prostasin with either Xenopus or rat ENaC in Xenopus oocytes resulted in a 60% to 80% increase in amiloride-sensitive sodium currents, and that the addition of aprotinin, a serine protease inhibitor, completely prevented this activation. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml mouse Prostasin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. We recommend that each mouse Prostasin standard solution and each sample is measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty