Would you like to log in again?
Human Granulysin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0756
76235-976EA 840 USD InStock
76235-976
Human Granulysin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Granulysin AccuSignal ELISA Kit
  • Natural and recombinant human Granulysin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: M1-L145
Granulysin is a substance released by cytotoxic T cells(CD8) when they are attached to infected body cells. The product of this gene is a member of the saposin-like protein(SAPLIP) family. It is mapped to 2p11.2. Granulysin functions to create holes in the target cell membrane and destroy it. It is able to induce apoptosis in target cells and also has antimicrobial action. This gene is expressed in cytolytic granules with perforin, a pore forming protein, and granzymes that are also involved in cytolysis. In addition to it, Granulysin is broadly antimicrobial, killing microbes that cause, for example, tuberculosis and malaria, and can destroy some tumors. A series of peptides generated from the amino acid sequence of Granulysin are potential antibiotics. It has been found that secretory Granulysin is a key molecule responsible for the disseminated keratinocyte death in SJS/TEN.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human Granulysin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Granulysin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty