Would you like to log in again?
EnzyFluo™ Collagen Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
ECOL-100
76236-120EA 614.66 USD InStock
76236-120
EnzyFluo™ Collagen Assay Kit, BioAssay Systems
Assays Cellular Assays
Collagen determination in biological and cosmetic products.
  • Safe, non-radioactive assay, fast and sensitive, convenient
  • Linear detection range (20 µl sample): 2 µg/ml to 50 µg/ml collagen in 96-well plate assay
  • High-throughput, "Add-mix-read" type assay
  • No wash and reagent transfer steps are involved
  • Can be readily automated for processing thousands of samples per day
  • Method: FL375/465 nm
Samples: Biological samples (example: plasma, serum, cells) and cosmetic samples (example: collagen solution). Collagen is the key structural protein of connective tissue and the most abundant protein in mammals. It occurs in many different types and forms with Types I -V being the most common. Aside from the crucial role it plays in the body, it has numerous medical applications such as its use in reconstructive surgery including bone and skin grafts.
It is also commonly used in cosmetics due to its anti-aging and skin healing properties. Assay methods available for quantifying collagen currently range from needing extensive hydrolysis procedures with acids and bases to using expensive antibodies and complicated protocols. Collagen assay kit delivers a very simple, safe, and sensitive method to quantify collagen in samples. In the first step of this procedure, collagen in the sample is enzymatically digested into peptides. Subsequently, the N-terminal glycine containing peptides react with the dye reagent to form a fluorescent complex. The fluorescence intensity of this product, measured at λex/em = 375/465 nm, is directly proportional to collagen concentration in the sample.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty