Would you like to log in again?
EnzyChrom™ Formate Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EFOR-100
76236-124EA 637.05 USD InStock
76236-124
EnzyChrom™ Formate Assay Kit, BioAssay Systems
Assays Cellular Assays
Direct determination of formate concentrations in biological (example: urine, blood, and more.) and other samples.
  • Fast and sensitive, convenient
  • Use of 10 µl sample
  • Linear detection range 0.050 to 5 mM Formate in 96-well plate assay
  • Room temperature assay, No 37 °C heater is needed
  • High-throughput, "Add-mix-read" type assay
  • Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: OD565 nm
The procedure involves adding a single working reagent, and reading the absorbance after 60 minutes. Formate (HCOO-) is the anion derived from formic acid, the simplest carboxylic acid. It is also the metabolic byproduct of formaldehyde metabolism in our body, and the eventual metabolic byproduct of methanol which is first broken down to formaldehyde.
At high levels, formate is neurotoxic to the central nervous system and can cause blindness, coma, and death. Although naturally present in the body at low levels, elevated levels of formate may be used as an indicator of formaldehyde exposure and methanol poisoning. Formate assay kit is based on formate dehydrogenase catalyzed oxidation of formate, which generates carbon dioxide and NADH that reduces a formazan (MTT) dye. The intensity of the reduced MTT, measured at 565 nm, is directly proportional to formate concentration in the sample. Samples: Urine, Serum and more.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty