Would you like to log in again?
QuantiChrom™ Aldehyde Dehydrogenase Inhibitor Screening Kit, BioAssay Systems
Supplier: BioAssay Systems
EIAL-100
76236-128EA 436.21 USD InStock
76236-128
QuantiChrom™ Aldehyde Dehydrogenase Inhibitor Screening Kit, BioAssay Systems
Assays Enzyme Assays
HTS for inhibitor screening and evaluation of aldehyde dehydrogenase inhibitors.
  • Rapid and reliable, can be completed in under an hour
  • Homogenous and convenient
  • "Mix-incubate-measure" type assay
  • No wash and reagent transfer steps are involved
  • Robust and amenable to HTS: can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: OD565 nm
  • Samples: aldehyde dehydrogenase
Aldehyde dehydrogenases (ALDHs) are a superfamily of oxidoreductases which catalyze the conversion of aldehydes to carboxylic acids. ALDH is crucial in the metabolism of alcohol as alcohol dehydrogenase breaks down ethanol to acetaldehyde. Acetaldehyde, which is toxic to the body, is in turn broken down by ALDH to acetic acid. Imbalances of aldehyde dehydrogenase have been linked to both alcoholism and alcohol sensitivity in people. Inhibitors of the enzyme have been used in cases to treat alcoholism in patients. Cancer stem cell populations also display a heightened activity of ALDH, making ALDH inhibition a promising anti-cancer therapy approach.
QuantiChrom™ Aldehyde Dehydrogenase Inhibitor Screening Kit is based on the enzymatic conversion of acetaldehyde to acetic acid and NADH by ALDH. The formed NADH in turn reduces a formazan reagent into a colored product the absorbance of which, measured at 565 nm, is proportional to the enzyme activity in the reaction. The percent inhibition of a test compound can be determined by comparing the activity of ALDH treated with a test compound to the activity of untreated ALDH.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty