Would you like to log in again?
EnzyChrom™ D-Mannitol Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EMNT-100
76236-140EA 659.81 USD InStock
76236-140
EnzyChrom™ D-Mannitol Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative determination of D-mannitol in biological, food, beverage and agriculture samples.
  • Fast and sensitive, convenient
  • Use of 20 μl sample
  • Linear detection range 0.007 to 3 mM mannitol in 96-well plate assay
  • Room temperature assay, no 37 °C heater is needed
  • High-throughput, "Add-mix-read" type assay
  • Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: OD565 nm
The procedure involves adding a single working reagent, and reading the absorbance after 30 minutes. D-Mannitol (C₆H₁₄O₆) is a sugar alcohol used in dietary supplement, sweetener, intestinal permeability test for leaky gut, etc. It also serves as a coating for hard candies, dried fruits, and chewing gums due to its low ability to attract and hold water molecules. In addition, it is an osmoprotectant for plants and is used clinically in osmotherapy to reduce intracranial pressure.
D-mannitol assay kit is based on mannitol dehydrogenase catalyzed oxidation of D-mannitol, which generates D-fructose and NADH that reduces a formazan (MTT) dye. The intensity of product color, measured at 565 nm is directly proportional to D-mannitol concentration in the sample. Samples: Biological, food, beverage and agricultural products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty