Would you like to log in again?
QuantiFluo™ Acid Phosphatase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
FACP-100
76236-142EA 226.66 USD InStock
76236-142
QuantiFluo™ Acid Phosphatase Assay Kit, BioAssay Systems
Assays Enzyme Assays
Acid Phosphatase activity determination in biological samples (example: plasma, serum, cell lysate, tissue samples.)
  • Fast and sensitive
  • Linear detection range (20 μl sample): 0.008 to 10 U/l for a 30 minute reaction
  • High-throughput, homogeneous "mix-incubate-measure" type assay
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day
  • Method: FL360/450 nm
  • Samples: Biological (example: serum) samples
Acid Phosphatase (ACP) is an enzyme which catalyzes the cleavage of phosphate groups from other molecules during digestion. Acid phosphatase can be found in lysosomes and become active after fusing with endosomes, acidifying the pH and, thus, creating an optimal environment for ACP. ACP can also be found in bone, spleen, liver, kidney and blood. Serum levels can be used as a biomarker for prostatic carcinoma, although prostate-specific antigen (PSA) is more widely used.
Non-radioactive, fluorimetric ACP assay is based on the cleavage of methylumbelliferyl phosphate. Methylumbelliferone becomes intensely fluorescent after addition of the stop reagent. The increase in fluorescence at 360/450 nm after addition of the stop reagent is directly proportional to the enzyme activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty