Would you like to log in again?
QuantiQuik™ Glucose Quick Test Strips, BioAssay Systems
Supplier: BioAssay Systems
QQGLUC10
76236-150EA 105.4 USD InStock
76236-150
QuantiQuik™ Glucose Quick Test Strips, BioAssay Systems
Assays Cellular Assays
Direct determination of glucose concentrations in food and beverage samples as well as biological (example: urine, blood, and more.) samples.
  • Fast and sensitive
  • Use of 20 or 100 µl sample
  • Semi-quantitative measurement between 0-720 mg/l (undiluted) Glucose
  • Convenient. No expensive lab equipment needed.
  • Sample treatment and assay can be performed in under 15 minutes
  • Method: Visual
  • Samples: Wine, beer, juice, and more
Glucose (C₆H₁₂O₆) is found in the food industry as either a natural component of the food or as an additive. Not only is glucose added to food and beverages for its impact on taste, texture, and color, but also because it can serve as a preservative.
QuantiQuik™ Glucose Test Strips are based on Glucose dehydrogenase catalyzed oxidation of glucose in which the formed NADH reduces a chromogenic reagent. The intensity of the product color is directly proportional to the glucose concentration in the sample.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty