Would you like to log in again?
QuantiQuik™ L-Lactic Acid Quick Test Strips, BioAssay Systems
Supplier: BioAssay Systems
QQLLAC10
76236-160EA 105.3 USD InStock
76236-160
QuantiQuik™ L-Lactic Acid Quick Test Strips, BioAssay Systems
Assays Cellular Assays
Direct determination of L-lactic acid concentrations in food and beverage samples as well as biological samples.
  • Fast and sensitive
  • Use of 20 or 100 µl sample
  • Semi-quantitative measurement between 0-360 mg/l (undiluted) L-lactic acid
  • Convenient, No expensive lab equipment needed
  • Sample treatment and assay can be performed in under 15 minutes
  • Method: Visual
  • Samples: Wine, beer, milk, and more
L-Lactic acid, or L-lactate, is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. L-lactic acid is added to many foods and beverages to give it a tart flavor. Increased levels of L-lactic acid in milk, egg, and fruit juices can be an indication of spoilage. In the wine industry, increasing levels of L-lactic acid and the decreasing levels of L-malic acid are monitored (Malolactic fermentation). In this process, the overall acidity of the wine is reduced and can lead to the improvement of the flavor of the wine.
QuantiQuik™ L-Lactic Acid Test Strips are based on L-lactate dehydrogenase catalyzed oxidation of L-lactate in which the formed NADH reduces a chromogenic reagent. The intensity of product color is directly proportional to L-lactate concentration in the sample.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty