Would you like to log in again?
Mouse CD48 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0787
76236-186EA 840 USD InStock
76236-186
Mouse CD48 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse CD48 AccuSignal ELISA Kit
  • Natural and recombinant mouse CD48.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: F23-S217
CD48 antigen(Cluster of Differentiation 48), also known as BLAST-1 or SLAMF2, is a protein that in humans is encoded by the CD48 gene. CD48 is a member of the CD2 subfamily of the immunoglobulin superfamily(IgSF) which includes SLAM(signaling lymphocyte activation molecules) proteins, such as CD84, CD150, CD229 and CD244. It is mapped to 1q23.3. CD48 is found on the surface of lymphocytes and other immune cells, dendritic cells and endothelial cells, and participates in activation and differentiation pathways in these cells. The encoded protein does not have a transmembrane domain, however, but is held at the cell surface by a GPI anchor via a C-terminal domain which maybe cleaved to yield a soluble form of the receptor.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse CD48 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD48 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty