Would you like to log in again?
Mouse MADCAM-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0801
76236-190EA 840 USD InStock
76236-190
Mouse MADCAM-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse MADCAM-1 AccuSignal ELISA Kit
  • Natural and recombinant mouse MADCAM-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q22-T365
MADCAM1(Mucosal Vascular Addressin Cell Adhesion Molecule 1), also known as MACAM1, is a protein that is encoded by the MADCAM1 gene. By PCR-based analysis of somatic cell hybrids, Leung et al(1997) mapped the MACAM1 gene to chromosome 19. The protein encoded by this gene is an endothelil cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1(alpha4 / beta7), L-selectin, and VLA-4(alpha4/beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin superfamily and is similar to ICAM-1 and VCAM-1.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml mouse MADCAM-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse MADCAM-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty