Would you like to log in again?
Human SDC1 - Syndecan-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0804
76236-196EA 636 USD InStock
76236-196
Human SDC1 - Syndecan-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human SDC1 - Syndecan-1 AccuSignal ELISA Kit
  • Natural and recombinant human Syndecan-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q23-G254
Syndecan 1, also known as SYND1 or CD138, is a protein which in humans is encoded by the SDC1 gene. The protein encoded by this gene is a transmembrane(type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. This gene is mapped to 2p24.1. The syndecans mediate cell binding, cell signaling, cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein. The syndecan 1 protein functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. What’s more, Syndecan-1 is also a sponge for growth factors, with binding largely via heparan sulfate chains.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human Syndecan-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human Syndecan-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty