Would you like to log in again?
Human ADAM9 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0806
76236-200EA 636 USD InStock
76236-200
Human ADAM9 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ADAM9 AccuSignal ELISA Kit
  • Natural and recombinant human ADAM9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A29-D697
Disintegrin and metalloproteinase domain-containing protein 9, also known as MCMP or MDC9 is an enzyme that in humans is encoded by the ADAM9 gene. This gene encodes a member of the ADAM('a disintegrin and metalloproteinase') family and exhibits a number of characteristics of class III reprolysins. This gene is mapped to 8p11.22. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human ADAM9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human ADAM9 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty