Would you like to log in again?
Human IL-34 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0826
76236-208EA 650 USD InStock
76236-208
Human IL-34 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Interleukin-34 AccuSignal ELISA Kit
  • Natural and recombinant human IL-34.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: N21-P242
Interleukin-34, or IL-34 is a protein belonging to a group of cytokines called interleukins. It is mapped to 16q22.1. The discovery of IL-34 protein in the red pulp of the spleen suggests involvement in growth and development of myeloid cells, consistent with its activity on monocytes. IL-34 is a recently discovered cytokine that acts on tissue resident macrophages and Langerhans cells upon binding the receptor for CSF-1, CSF-1R. It also can promote the differentiation and viability of monocytes and macrophages through the colony-stimulating factor-1 receptor. In addition, it promotes the development, survival, and function of microglia and Langerhans cells, therefore, this cytokine may predominately function in brain and skin biology.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human IL-34 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-34 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty