Would you like to log in again?
Human ANGPTL3 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0833
76236-222EA 636 USD InStock
76236-222
Human ANGPTL3 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ANGPTL3 AccuSignal ELISA Kit
  • Natural and recombinant human ANGPTL3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: S17-E460
ANGPTL3, also known as Angiopoietin-like 3 or HYPLl, is a protein that in humans is encoded by the ANGPTL3 gene. It is mapped to 1p31.3. The protein encoded by this gene is a member of the angiopoietin-like family of secreted factors. It is expressed predominantly in the liver, and has the characteristic structure of angiopoietins, consisting of a signal peptide, N-terminal coiled-coil domain, and the C-terminal fibrinogen (FBN)-like domain. ANGPTL3 also acts as dual inhibitor of lipoprotein lipase (LPL) and endothelial lipase (EL), and increases plasma triglyceride and HDL cholesterol in rodents. It can inhibit endothelial lipase to catalyze HDL-phospholipid and increase HDL-PL levels.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml human ANGPTL3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human ANGPTL3 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty